Learn More
Methyl 6-chloropyridine-2-carboxylate, 95%, Thermo Scientific Chemicals
$82.47 - $306.04
Chemical Identifiers
CAS | 6636-55-1 |
---|---|
Molecular Formula | C7H6ClNO2 |
Molecular Weight (g/mol) | 171.58 |
MDL Number | MFCD00155391 |
InChI Key | TWUXBVMXSBEKHA-UHFFFAOYSA-N |
Synonym | methyl 6-chloropicolinate, methyl 6-chloro-2-pyridinecarboxylate, 2-pyridinecarboxylic acid, 6-chloro-, methyl ester, 2-chloropyridine-6-carboxylate methyl, 6-chloro-2-picolinic acid methyl ester, 6-chloropicolinic acid methyl ester, methyl 2-chloro-6-pyridinecarboxylate, methyl 2-chloropyridine-6-carboxylate, 6-chloro-pyridine-2-carboxylic acid methyl ester, 6-chloro-2-pyridinecarboxylic acid methyl ester |
PubChem CID | 242723 |
IUPAC Name | methyl 6-chloropyridine-2-carboxylate |
SMILES | COC(=O)C1=NC(=CC=C1)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6402306
|
Thermo Scientific Chemicals
H6402306 |
5 g |
Each for $82.47
|
|
|||||
AAH6402314
|
Thermo Scientific Chemicals
H6402314 |
25 g |
Each for $306.04
|
|
|||||
Description
Methyl 6-chloropyridine-2-carboxylate was coupled with 2-(trimethylstannyl)pyridine to afford methyl 2,2?-bipyridine-6-carboxylate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 6-chloropyridine-2-carboxylate was coupled with 2-(trimethylstannyl)pyridine to afford methyl 2,2′-bipyridine-6-carboxylate.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
Chemical Identifiers
6636-55-1 | |
171.58 | |
TWUXBVMXSBEKHA-UHFFFAOYSA-N | |
242723 | |
COC(=O)C1=NC(=CC=C1)Cl |
C7H6ClNO2 | |
MFCD00155391 | |
methyl 6-chloropicolinate, methyl 6-chloro-2-pyridinecarboxylate, 2-pyridinecarboxylic acid, 6-chloro-, methyl ester, 2-chloropyridine-6-carboxylate methyl, 6-chloro-2-picolinic acid methyl ester, 6-chloropicolinic acid methyl ester, methyl 2-chloro-6-pyridinecarboxylate, methyl 2-chloropyridine-6-carboxylate, 6-chloro-pyridine-2-carboxylic acid methyl ester, 6-chloro-2-pyridinecarboxylic acid methyl ester | |
methyl 6-chloropyridine-2-carboxylate |
Specifications
6636-55-1 | |
Yellow | |
MFCD00155391 | |
128580 | |
Slightly soluble in water. | |
COC(=O)C1=NC(=CC=C1)Cl | |
171.58 | |
171.58 | |
Solid |
92°C to 98°C (literature) | |
C7H6ClNO2 | |
5 g | |
methyl 6-chloropicolinate, methyl 6-chloro-2-pyridinecarboxylate, 2-pyridinecarboxylic acid, 6-chloro-, methyl ester, 2-chloropyridine-6-carboxylate methyl, 6-chloro-2-picolinic acid methyl ester, 6-chloropicolinic acid methyl ester, methyl 2-chloro-6-pyridinecarboxylate, methyl 2-chloropyridine-6-carboxylate, 6-chloro-pyridine-2-carboxylic acid methyl ester, 6-chloro-2-pyridinecarboxylic acid methyl ester | |
TWUXBVMXSBEKHA-UHFFFAOYSA-N | |
methyl 6-chloropyridine-2-carboxylate | |
242723 | |
95% | |
Methyl 6-chloropyridine-2-carboxylate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only