Learn More
Methyl acetate, 99%, extra pure
CAS: 79-20-9 | C3H6O2 | 74.08 g/mol
$169.27 - $264.39
Chemical Identifiers
| CAS | 79-20-9 |
|---|---|
| Molecular Formula | C3H6O2 |
| Molecular Weight (g/mol) | 74.08 |
| MDL Number | MFCD00008711 |
| InChI Key | KXKVLQRXCPHEJC-UHFFFAOYSA-N |
| Synonym | tereton, devoton, acetic acid, methyl ester, methyl ethanoate, acetic acid methyl ester, methylacetat, acetate de methyle, methylacetaat, methyl acetic ester, octan metylu |
| PubChem CID | 6584 |
| ChEBI | CHEBI:77700 |
| IUPAC Name | methyl acetate |
| SMILES | CC(=O)OC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC181380010
|
Thermo Scientific Chemicals
181380010 |
1 L | Glass Bottle |
Each for $169.27
Case of 4 Each for $0.00
|
|
||||
|
AC181380025
|
Thermo Scientific Chemicals
181380025 |
2.5 L | Glass Bottle |
Each for $264.39
Case of 4 Each for $0.00
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 79-20-9 | |
| 74.08 | |
| KXKVLQRXCPHEJC-UHFFFAOYSA-N | |
| 6584 | |
| methyl acetate |
| C3H6O2 | |
| MFCD00008711 | |
| tereton, devoton, acetic acid, methyl ester, methyl ethanoate, acetic acid methyl ester, methylacetat, acetate de methyle, methylacetaat, methyl acetic ester, octan metylu | |
| CHEBI:77700 | |
| CC(=O)OC |
Specifications
| 79-20-9 | |
| 0.9300g/mL | |
| −10°C | |
| 98.5% min. (GC) | |
| C3H6O2 | |
| CH3CO2CH3 | |
| 1 L | |
| 0.001% max. | |
| 15, 6082 | |
| Solubility in water: 250g/L (20°C). Other solubilities: miscible with alcohol and ether | |
| CC(=O)OC | |
| 74.08 | |
| 0.38 mPa.s (20°C) | |
| 74.08 | |
| Extra Pure | |
| Methyl acetate |
| -98.0°C | |
| 57.4°C | |
| Authentic | |
| Glass Bottle | |
| 1.3600 to 1.3620 | |
| MFCD00008711 | |
| 02, 224 | |
| 0.93 | |
| tereton, devoton, acetic acid, methyl ester, methyl ethanoate, acetic acid methyl ester, methylacetat, acetate de methyle, methylacetaat, methyl acetic ester, octan metylu | |
| KXKVLQRXCPHEJC-UHFFFAOYSA-N | |
| methyl acetate | |
| 6584 | |
| CHEBI:77700 | |
| 99% | |
| Liquid |
Safety and Handling
GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 201-185-2
RUO – Research Use Only