missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl Acetate, Technical, Approx. 75%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor M1230200LTBL

 View more versions of this product

Catalog No. 18611668

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

79-20-9
Drum
∼75%
1
200 L
Technical

Chemical Identifiers

79-20-9
Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food use. Read and understand the label and Safety Data Sheet (SDS) prior to use.