missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl acetoacetate, 99+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 126150025

Catalog No. AC126150025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Methyl acetoacetate
1.0770g/mL
67°C
99% min. (GC)
C5H8O3
CH3COCH2CO2CH3
2.5L
13,60
15, 6083
Solubility in water: 460g/L (20°C). Other solubilities: miscible with alcohol and ether,soluble in carbon tetrachloride
COC(=O)CC(C)=O
116.12
1.8 mPa.s (20°C)
99+%
105-45-3
169.0°C to 170.0°C
Authentic
Glass bottle
1.4175 to 1.4295
MFCD00008784
03, 632
1.077
methyl acetoacetate, acetoacetic acid methyl ester, methylacetoacetate, methyl 3-oxobutyrate, butanoic acid, 3-oxo-, methyl ester, methyl acetylacetate, methyl acetylacetonate, 3-oxobutanoic acid methyl ester, acetoacetic methyl ester, acetoacetic acid, methyl ester
WRQNANDWMGAFTP-UHFFFAOYSA-N
methyl 3-oxobutanoate
7757
116.12
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 203-299-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only