Learn More
Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 126190010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl acrylate | |
96-33-3,150-76-5 | |
0.9560g/mL | |
-3°C | |
98.5% min. (GC) | |
C4H6O2 | |
H2C=CHCO2CH3 | |
1 L | |
05,439; 13,183; 17,183 | |
15, 6085 | |
Solubility in water: 60g/L (20°C). Other solubilities: soluble in alcohol and ether | |
COC(=O)C=C | |
86.09 | |
7294 | |
CHEBI:82482 | |
99% |
Stabilized | |
-75°C | |
80°C | |
Authentic | |
Glass bottle | |
1.4010 to 1.404 | |
MFCD00008627 | |
02, 399 | |
0.956 | |
methyl acrylate, methylacrylate, acrylic acid methyl ester, 2-propenoic acid, methyl ester, methyl 2-propenoate, methyl propenoate, methylacrylaat, metilacrilato, methyl-acrylat, methoxycarbonylethylene | |
BAPJBEWLBFYGME-UHFFFAOYSA-N | |
methyl prop-2-enoate | |
10 to 20ppm MEHQ | |
0.50 mPa.s (20°C) | |
86.09 | |
Liquid |
Chemical Identifiers
96-33-3 | |
86.09 | |
BAPJBEWLBFYGME-UHFFFAOYSA-N | |
7294 | |
methyl prop-2-enoate |
C4H6O2 | |
MFCD00008627 | |
methyl acrylate, methylacrylate, acrylic acid methyl ester, 2-propenoic acid, methyl ester, methyl 2-propenoate, methyl propenoate, methylacrylaat, metilacrilato, methyl-acrylat, methoxycarbonylethylene | |
CHEBI:82482 | |
COC(=O)C=C |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Toxic if inhaled
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Ground/Bond container and receiving equipment.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a positi
GHS Signal Word: Danger
EINECSNumber : 202-500-6
RUO – Research Use Only