missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 126195000

 View more versions of this product

Catalog No. AC126195000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methyl acrylate
96-33-3,150-76-5
0.9560g/mL
-3°C
98.5% min. (GC)
C4H6O2
H2C=CHCO2CH3
500 mL
05,439; 13,183; 17,183
15, 6085
Solubility in water: 60g/L (20°C). Other solubilities: soluble in alcohol and ether
COC(=O)C=C
86.09
7294
CHEBI:82482
99%
Stabilized
-75°C
80°C
Authentic
Glass bottle
1.4010 to 1.404
MFCD00008627
02, 399
0.956
methyl acrylate, methylacrylate, acrylic acid methyl ester, 2-propenoic acid, methyl ester, methyl 2-propenoate, methyl propenoate, methylacrylaat, metilacrilato, methyl-acrylat, methoxycarbonylethylene
BAPJBEWLBFYGME-UHFFFAOYSA-N
methyl prop-2-enoate
10 to 20ppm MEHQ
0.50 mPa.s (20°C)
86.09
Liquid

Chemical Identifiers

96-33-3
86.09
BAPJBEWLBFYGME-UHFFFAOYSA-N
7294
methyl prop-2-enoate
C4H6O2
MFCD00008627
methyl acrylate, methylacrylate, acrylic acid methyl ester, 2-propenoic acid, methyl ester, methyl 2-propenoate, methyl propenoate, methylacrylaat, metilacrilato, methyl-acrylat, methoxycarbonylethylene
CHEBI:82482
COC(=O)C=C
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Toxic if inhaled

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Ground/Bond container and receiving equipment.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a positi

GHS Signal Word: Danger

EINECSNumber : 202-500-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only