missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl Acrylate (stabilized with MEHQ) 99.0+%, TCI America™
$39.51 - $55.30
Chemical Identifiers
CAS | 96-33-3 |
---|---|
Molecular Formula | C4H6O2 |
Molecular Weight (g/mol) | 86.09 |
MDL Number | MFCD00008627 |
InChI Key | BAPJBEWLBFYGME-UHFFFAOYSA-N |
Synonym | methyl acrylate, methylacrylate, acrylic acid methyl ester, 2-propenoic acid, methyl ester, methyl 2-propenoate, methyl propenoate, methylacrylaat, metilacrilato, methyl-acrylat, methoxycarbonylethylene |
PubChem CID | 7294 |
ChEBI | CHEBI:82482 |
IUPAC Name | methyl prop-2-enoate |
SMILES | COC(=O)C=C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
A014525ML
|
TCI America
A014525ML |
25 mL |
Each for $39.51
|
|
|||||
A0145500ML
|
TCI America
A0145500ML |
500 mL |
Each for $55.30
|
|
|||||
Chemical Identifiers
96-33-3 | |
86.09 | |
BAPJBEWLBFYGME-UHFFFAOYSA-N | |
7294 | |
methyl prop-2-enoate |
C4H6O2 | |
MFCD00008627 | |
methyl acrylate, methylacrylate, acrylic acid methyl ester, 2-propenoic acid, methyl ester, methyl 2-propenoate, methyl propenoate, methylacrylaat, metilacrilato, methyl-acrylat, methoxycarbonylethylene | |
CHEBI:82482 | |
COC(=O)C=C |
Specifications
96-33-3 | |
Colorless | |
C4H6O2 | |
25 mL | |
methyl acrylate, methylacrylate, acrylic acid methyl ester, 2-propenoic acid, methyl ester, methyl 2-propenoate, methyl propenoate, methylacrylaat, metilacrilato, methyl-acrylat, methoxycarbonylethylene | |
COC(=O)C=C | |
86.09 | |
CHEBI:82482 | |
≥99.0% (GC) | |
Methyl Acrylate (stabilized with MEHQ) |
-75°C | |
80°C | |
MFCD00008627 | |
1919 | |
BAPJBEWLBFYGME-UHFFFAOYSA-N | |
methyl prop-2-enoate | |
7294 | |
86.09 | |
Liquid |
Safety and Handling
EINECSNumber : (2)-0987
RTECSNumber : AT2800000
TSCA : Yes