missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl benzoate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A124040B

 View more versions of this product

Catalog No. AAA124040B


Explore available promotions

Description

Description

Methyl benzoate is used in perfumery. It also finds use as a solvent. It acts as an intermediate and odor agents. Further, it is used to attract insects such as orchid bees. It is also used for cellulose esters, cellulose ethers, synthetic resin and rubber solvent and polyester fibers to assist in the preparation of flavor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl benzoate is used in perfumery. It also finds use as a solvent. It acts as an intermediate and odor agents. Further, it is used to attract insects such as orchid bees. It is also used for cellulose esters, cellulose ethers, synthetic resin and rubber solvent and polyester fibers to assist in the preparation of flavor.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications

Specifications

Methyl benzoate
-12°C
198°C to 200°C
99%
1.517
1000 g
14,6024
Miscible with water.
COC(=O)C1=CC=CC=C1
136.15
CHEBI:72775
99%
93-58-3
1.094
82°C (179°F)
C8H8O2
MFCD00008421
1072099
methylbenzoate, benzoic acid, methyl ester, clorius, benzoic acid methyl ester, niobe oil, oil of niobe, methyl benzenecarboxylate, essence of niobe, oniobe oil, oxidate le
QPJVMBTYPHYUOC-UHFFFAOYSA-N
methyl benzoate
7150
136.15
Liquid

Chemical Identifiers

93-58-3
136.15
QPJVMBTYPHYUOC-UHFFFAOYSA-N
7150
methyl benzoate
C8H8O2
MFCD00008421
methylbenzoate, benzoic acid, methyl ester, clorius, benzoic acid methyl ester, niobe oil, oil of niobe, methyl benzenecarboxylate, essence of niobe, oniobe oil, oxidate le
CHEBI:72775
COC(=O)C1=CC=CC=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.

P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c

H227-H302

EINECSNumber : 202-259-7

RTECSNumber : DH3850000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only