missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl-beta-cyclodextrin (mixture of several Methylated), TCI America™
$215.27 - $1,074.40
Chemical Identifiers
CAS | 128446-36-6 |
---|---|
Molecular Formula | C54H94O35 |
Molecular Weight (g/mol) | 1303.311 |
MDL Number | MFCD00074980 |
InChI Key | YZOUYRAONFXZSI-SBHWVFSVSA-N |
Synonym | beta-cyclodextrin methyl ethers, beta-cyd, methyl-?cyclodextrin, beta-w7 m1.8, dimethyl-, a-cyclodextrin, .beta.-cyclodextrin,methylethers, methyl-b-cyclodextrin cell culture*tested |
PubChem CID | 51051622 |
SMILES | COC1C(C2C(OC1OC3C(OC(C(C3OC)OC)OC4C(OC(C(C4OC)OC)OC5C(OC(C(C5OC)OC)OC6C(OC(C(C6OC)OC)OC7C(OC(C(C7OC)OC)OC8C(OC(O2)C(C8O)OC)CO)CO)CO)CO)CO)CO)CO)O |
Chemical Identifiers
128446-36-6 | |
1303.311 | |
YZOUYRAONFXZSI-SBHWVFSVSA-N | |
51051622 |
C54H94O35 | |
MFCD00074980 | |
beta-cyclodextrin methyl ethers, beta-cyd, methyl-?cyclodextrin, beta-w7 m1.8, dimethyl-, a-cyclodextrin, .beta.-cyclodextrin,methylethers, methyl-b-cyclodextrin cell culture*tested | |
COC1C(C2C(OC1OC3C(OC(C(C3OC)OC)OC4C(OC(C(C4OC)OC)OC5C(OC(C(C5OC)OC)OC6C(OC(C(C6OC)OC)OC7C(OC(C(C7OC)OC)OC8C(OC(O2)C(C8O)OC)CO)CO)CO)CO)CO)CO)CO)O |
Specifications
128446-36-6 | |
MFCD00074980 | |
beta-cyclodextrin methyl ethers, beta-cyd, methyl-?cyclodextrin, beta-w7 m1.8, dimethyl-, a-cyclodextrin, .beta.-cyclodextrin,methylethers, methyl-b-cyclodextrin cell culture*tested | |
COC1C(C2C(OC1OC3C(OC(C(C3OC)OC)OC4C(OC(C(C4OC)OC)OC5C(OC(C(C5OC)OC)OC6C(OC(C(C6OC)OC)OC7C(OC(C(C7OC)OC)OC8C(OC(O2)C(C8O)OC)CO)CO)CO)CO)CO)CO)CO)O | |
51051622 | |
Methyl-beta-cyclodextrin (mixture of several Methylated) |
C54H94O35 | |
25 g | |
YZOUYRAONFXZSI-SBHWVFSVSA-N | |
1303.311 | |
Crystalline Powder |
Safety and Handling
TSCA : Yes