missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl beta-L-Fucopyranoside, TCI America™
Supplier: TCI America M10501G
Specifications
Methyl beta-L-Fucopyranoside | |
123°C | |
C7H14O5 | |
1 g | |
CC1C(C(C(C(O1)OC)O)O)O | |
178.184 | |
178.18 |
24332-98-7 | |
White | |
MFCD00069803 | |
OHWCAVRRXKJCRB-XUVCUMPTSA-N | |
(2S,3S,4R,5S,6S)-2-methoxy-6-methyloxane-3,4,5-triol | |
7167925 | |
Crystalline Powder |
Chemical Identifiers
24332-98-7 | |
178.184 | |
OHWCAVRRXKJCRB-XUVCUMPTSA-N | |
(2S,3S,4R,5S,6S)-2-methoxy-6-methyloxane-3,4,5-triol |
C7H14O5 | |
MFCD00069803 | |
7167925 | |
CC1C(C(C(C(O1)OC)O)O)O |
Safety and Handling
TSCA : No