missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl biphenyl-4-carboxylate, 98+%, Thermo Scientific Chemicals
$71.18 - $361.38
Chemical Identifiers
CAS | 720-75-2 |
---|---|
Molecular Formula | C14H12O2 |
Molecular Weight (g/mol) | 212.25 |
MDL Number | MFCD00017200 |
InChI Key | GATUGNVDXMYTJX-UHFFFAOYSA-N |
Synonym | methyl 4-biphenylcarboxylate, methyl biphenyl-4-carboxylate, methyl 1,1'-biphenyl-4-carboxylate, methyl p-phenylbenzoate, 4-biphenylcarboxylic acid, methyl ester, 1,1'-biphenyl-4-carboxylic acid, methyl ester, p-phenylbenzoic acid methyl ester, methyl-4-phenylbenzoate, 4-phenylbenzoic acid methyl ester |
PubChem CID | 69757 |
IUPAC Name | methyl 4-phenylbenzoate |
SMILES | COC(=O)C1=CC=C(C=C1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1343006
|
Thermo Scientific Chemicals
A1343006 |
5 g |
Each for $71.18
|
|
|||||
AAA1343018
|
Thermo Scientific Chemicals
A1343018 |
50 g |
Each for $361.38
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
720-75-2 | |
212.25 | |
GATUGNVDXMYTJX-UHFFFAOYSA-N | |
69757 | |
COC(=O)C1=CC=C(C=C1)C1=CC=CC=C1 |
C14H12O2 | |
MFCD00017200 | |
methyl 4-biphenylcarboxylate, methyl biphenyl-4-carboxylate, methyl 1,1'-biphenyl-4-carboxylate, methyl p-phenylbenzoate, 4-biphenylcarboxylic acid, methyl ester, 1,1'-biphenyl-4-carboxylic acid, methyl ester, p-phenylbenzoic acid methyl ester, methyl-4-phenylbenzoate, 4-phenylbenzoic acid methyl ester | |
methyl 4-phenylbenzoate |
Specifications
720-75-2 | |
Odorless | |
MFCD00017200 | |
1954046 | |
GATUGNVDXMYTJX-UHFFFAOYSA-N | |
methyl 4-phenylbenzoate | |
69757 | |
≥98% |
116°C to 119°C | |
C14H12O2 | |
5 g | |
methyl 4-biphenylcarboxylate, methyl biphenyl-4-carboxylate, methyl 1,1'-biphenyl-4-carboxylate, methyl p-phenylbenzoate, 4-biphenylcarboxylic acid, methyl ester, 1,1'-biphenyl-4-carboxylic acid, methyl ester, p-phenylbenzoic acid methyl ester, methyl-4-phenylbenzoate, 4-phenylbenzoic acid methyl ester | |
COC(=O)C1=CC=C(C=C1)C1=CC=CC=C1 | |
212.25 | |
212.25 | |
Methyl biphenyl-4-carboxylate |
Safety and Handling
EINECSNumber : 211-954-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only