missing translation for 'onlineSavingsMsg'
Learn More

Methyl bromoacetate, 98+%

CAS: 96-32-2 | C3H5BrO2 | 152.975 g/mol

$88.99 - $681.41

Chemical Identifiers

CAS 96-32-2
Molecular Formula C3H5BrO2
Molecular Weight (g/mol) 152.975
MDL Number MFCD00000189
InChI Key YDCHPLOFQATIDS-UHFFFAOYSA-N
Synonym methyl bromoacetate, bromoacetic acid methyl ester, methyl monobromoacetate, acetic acid, bromo-, methyl ester, methyl alpha-bromoacetate, methylbromoacetate, acetic acid, 2-bromo-, methyl ester, methylester kyseliny bromoctove, methyl .alpha.-bromoacetate, methylester kyseliny bromoctove czech
PubChem CID 60984
IUPAC Name methyl 2-bromoacetate
SMILES COC(=O)CBr
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1060518
View Documents
Thermo Scientific Chemicals
A1060518
50 g
Each for $88.99
Only null left
 
AAA1060530
View Documents
Thermo Scientific Chemicals
A1060530
250 g
Each for $227.73
Only null left
 
AAA106050B
View Documents
Thermo Scientific Chemicals
A106050B
1000 g
Each for $681.41
Only null left
 
Description

Description

Methyl bromoacetate is used in the synthesis of coumarins and cis-cyclopropanes. It reacts with the conjugate base of (methylmethoxycarbene)pentacarbonylchromium(0) to prepare alkylated carbene complexes. Further, it is used to make vitamins and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl bromoacetate is used in the synthesis of coumarins and cis-cyclopropanes. It reacts with the conjugate base of (methylmethoxycarbene)pentacarbonylchromium(0) to prepare alkylated carbene complexes. Further, it is used to make vitamins and pharmaceuticals.

Solubility
Miscible with methanol, ether and acetone. Slightly miscible with water

Notes
Incompatible with acids, bases, oxidizing agents and reducing agents.
Specifications

Chemical Identifiers

96-32-2
152.975
YDCHPLOFQATIDS-UHFFFAOYSA-N
60984
COC(=O)CBr
C3H5BrO2
MFCD00000189
methyl bromoacetate, bromoacetic acid methyl ester, methyl monobromoacetate, acetic acid, bromo-, methyl ester, methyl alpha-bromoacetate, methylbromoacetate, acetic acid, 2-bromo-, methyl ester, methylester kyseliny bromoctove, methyl .alpha.-bromoacetate, methylester kyseliny bromoctove czech
methyl 2-bromoacetate

Specifications

96-32-2
1.655
62°C (143°F)
C3H5BrO2
BrCH2CO2CH3
50 g
506256
Miscible with methanol,ether and acetone. Slightly miscible with water
COC(=O)CBr
152.975
152.98
Methyl bromoacetate
-50°C
144°C to 145°C
Pungent
1.4585
MFCD00000189
UN2643
methyl bromoacetate, bromoacetic acid methyl ester, methyl monobromoacetate, acetic acid, bromo-, methyl ester, methyl alpha-bromoacetate, methylbromoacetate, acetic acid, 2-bromo-, methyl ester, methylester kyseliny bromoctove, methyl .alpha.-bromoacetate, methylester kyseliny bromoctove czech
YDCHPLOFQATIDS-UHFFFAOYSA-N
methyl 2-bromoacetate
60984
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H314-H318-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P235-P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c

H227-H301-H314-H317-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METHYL BROMOACETATE

EINECSNumber : 202-499-2

RTECSNumber : AF6300000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only