missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl cinnamate, 98%
CAS: 103-26-4 | C10H10O2 | 162.19 g/mol
$57.74 - $190.46
Chemical Identifiers
| CAS | 103-26-4 |
|---|---|
| Molecular Formula | C10H10O2 |
| Molecular Weight (g/mol) | 162.19 |
| MDL Number | MFCD00008458 |
| InChI Key | CCRCUPLGCSFEDV-BQYQJAHWSA-N |
| Synonym | methyl cinnamate, methyl trans-cinnamate, methyl e-cinnamate, cinnamic acid methyl ester, methyl cinnamylate, methyl 3-phenylacrylate, methyl e-3-phenylprop-2-enoate, methyl 3-phenylpropenoate, trans-cinnamic acid methyl ester, trans-methyl cinnamate |
| PubChem CID | 637520 |
| ChEBI | CHEBI:6857 |
| IUPAC Name | methyl (E)-3-phenylprop-2-enoate |
| SMILES | COC(=O)C=CC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC279760050
|
Thermo Scientific Chemicals
279760050 |
5 g | Glass bottle |
Each for $57.74
|
|
||||
|
AC279765000
|
Thermo Scientific Chemicals
279765000 |
500 g | Glass bottle |
Each for $190.46
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 103-26-4 | |
| 162.19 | |
| CCRCUPLGCSFEDV-BQYQJAHWSA-N | |
| 637520 | |
| methyl (E)-3-phenylprop-2-enoate |
| C10H10O2 | |
| MFCD00008458 | |
| methyl cinnamate, methyl trans-cinnamate, methyl e-cinnamate, cinnamic acid methyl ester, methyl cinnamylate, methyl 3-phenylacrylate, methyl e-3-phenylprop-2-enoate, methyl 3-phenylpropenoate, trans-cinnamic acid methyl ester, trans-methyl cinnamate | |
| CHEBI:6857 | |
| COC(=O)C=CC1=CC=CC=C1 |
Specifications
| 103-26-4 | |
| 33.5°C to 36°C | |
| 260°C to 262°C | |
| Authentic | |
| Glass bottle | |
| C6H5CH=CHCO2CH3 | |
| 5 g | |
| 15, 2299 | |
| Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,glycerol,most fixed,oils and mineral oil | |
| COC(=O)C=CC1=CC=CC=C1 | |
| 162.19 | |
| CHEBI:6857 | |
| 98% | |
| Methyl cinnamate, 98% |
| 1mg KOH/g max. | |
| White to Yellow | |
| >110°C | |
| 97.5% min. (GC) | |
| C10H10O2 | |
| MFCD00008458 | |
| 09, 581 | |
| methyl cinnamate, methyl trans-cinnamate, methyl e-cinnamate, cinnamic acid methyl ester, methyl cinnamylate, methyl 3-phenylacrylate, methyl e-3-phenylprop-2-enoate, methyl 3-phenylpropenoate, trans-cinnamic acid methyl ester, trans-methyl cinnamate | |
| CCRCUPLGCSFEDV-BQYQJAHWSA-N | |
| methyl (E)-3-phenylprop-2-enoate | |
| 637520 | |
| 162.19 | |
| Fused Crystalline Mass |
Safety and Handling
EINECSNumber : 203-093-8
RTECSNumber : GE0190000
TSCA : TSCA
RUO – Research Use Only