Learn More
Methyl decanoate, 98+%, Thermo Scientific Chemicals
$85.47 - $306.79
Chemical Identifiers
CAS | 110-42-9 |
---|---|
Molecular Formula | C11H22O2 |
Molecular Weight (g/mol) | 186.30 |
MDL Number | MFCD00009580 |
InChI Key | YRHYCMZPEVDGFQ-UHFFFAOYSA-N |
Synonym | methyl caprate, decanoic acid, methyl ester, methyl caprinate, capric acid methyl ester, decanoic acid methyl ester, methyl-n-caprate, uniphat a30, metholene 2095, methyl n-decanoate, methyl n-caprate |
PubChem CID | 8050 |
IUPAC Name | methyl decanoate |
SMILES | CCCCCCCCCC(=O)OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1565818
|
Thermo Scientific Chemicals
A1565818 |
50 g |
Each for $85.47
|
|
|||||
AAA1565822
|
Thermo Scientific Chemicals
A1565822 |
100 g |
Each for $141.53
|
|
|||||
AAA1565830
|
Thermo Scientific Chemicals
A1565830 |
250 g |
Each for $306.79
|
|
|||||
Description
It is employed as an intermediate for caproic acid, detergents, emulsifiers, wetting agents, stabilizers, resins, lubricants, plasticizers and flavoring. As a chemical intermediate for alkanolamides, fatty alcohols & acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as an intermediate for caproic acid, detergents, emulsifiers, wetting agents, stabilizers, resins, lubricants, plasticizers and flavoring. As a chemical intermediate for alkanolamides, fatty alcohols & acids.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
110-42-9 | |
186.30 | |
YRHYCMZPEVDGFQ-UHFFFAOYSA-N | |
8050 | |
CCCCCCCCCC(=O)OC |
C11H22O2 | |
MFCD00009580 | |
methyl caprate, decanoic acid, methyl ester, methyl caprinate, capric acid methyl ester, decanoic acid methyl ester, methyl-n-caprate, uniphat a30, metholene 2095, methyl n-decanoate, methyl n-caprate | |
methyl decanoate |
Specifications
110-42-9 | |
0.877 | |
94°C (201°F) | |
1.425 | |
50 g | |
methyl caprate, decanoic acid, methyl ester, methyl caprinate, capric acid methyl ester, decanoic acid methyl ester, methyl-n-caprate, uniphat a30, metholene 2095, methyl n-decanoate, methyl n-caprate | |
YRHYCMZPEVDGFQ-UHFFFAOYSA-N | |
methyl decanoate | |
8050 | |
99% |
-18°C | |
223°C to 225°C | |
C11H22O2 | |
MFCD00009580 | |
1759170 | |
Soluble in alcohol. Insoluble in water. | |
CCCCCCCCCC(=O)OC | |
186.30 | |
186.3 | |
Methyl decanoate |
Safety and Handling
P210-P233-P240-P241-P242-P243-P280-P374-P501c
H227
EINECSNumber : 203-766-6
RTECSNumber : HD9830000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only