missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl dichloroacetate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2439936

 View more versions of this product

Catalog No. AAB2439936


Explore available promotions

Description

Description

Methyl dichloroacetate may be used in the diastereoselctive synthesis of bicyclic chlorocyclopropane, as difunctional initiator during atom transfer radical polymerization of methyl or n-butyl acrylate, as reagent during CrCl2-induced olefination of aldehydes. It is a reactant involved in nucleophilic substitution reactions with nitro(pentafluorosulfanyl)benzenes, azide-alkyl halide cycloaddition and atom transfer radical addition, Chemical modification of STn antigen for enhancement of synthetic carbohydrate vaccines, three component reaction of arynes with cyclic ethers and active methines, catalytic radical haloalkylation for synthesis of neodysidenin and addition reactions for syntehsis of enantiopure 3,4-diamino esters or amides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl dichloroacetate may be used in the diastereoselctive synthesis of bicyclic chlorocyclopropane, as difunctional initiator during atom transfer radical polymerization of methyl or n-butyl acrylate, as reagent during CrCl2-induced olefination of aldehydes. It is a reactant involved in nucleophilic substitution reactions with nitro(pentafluorosulfanyl)benzenes, azide-alkyl halide cycloaddition and atom transfer radical addition, Chemical modification of STn antigen for enhancement of synthetic carbohydrate vaccines, three component reaction of arynes with cyclic ethers and active methines, catalytic radical haloalkylation for synthesis of neodysidenin and addition reactions for syntehsis of enantiopure 3,4-diamino esters or amides.

Solubility
Soluble in water. (4 g/L).

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure proper ventilation.
Specifications

Specifications

Methyl dichloroacetate
-52°C
142°C to 144°C
Pungent
1.442
MFCD00000843
UN2299
Soluble in water. (4g/L).
COC(=O)C(Cl)Cl
142.963
142.97
116-54-1
1.38
70°C (158°F)
C3H4Cl2O2
Cl2CHCO2CH3
500 g
methyl dichloroacetate, dichloroacetic acid methyl ester, acetic acid, dichloro-, methyl ester, ccris 7740, acetic acid, 2,2-dichloro-, methyl ester, methyldichloroethanoate, acmc-1brpm, dichloroacetic acid methyl, methyl2,2-dichloroacetate, wln: gygvo1
HKMLRUAPIDAGIE-UHFFFAOYSA-N
methyl 2,2-dichloroacetate
8315
99%

Chemical Identifiers

116-54-1
142.963
HKMLRUAPIDAGIE-UHFFFAOYSA-N
8315
COC(=O)C(Cl)Cl
C3H4Cl2O2
MFCD00000843
methyl dichloroacetate, dichloroacetic acid methyl ester, acetic acid, dichloro-, methyl ester, ccris 7740, acetic acid, 2,2-dichloro-, methyl ester, methyldichloroethanoate, acmc-1brpm, dichloroacetic acid methyl, methyl2,2-dichloroacetate, wln: gygvo1
methyl 2,2-dichloroacetate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H227-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: METHYL DICHLOROACETATE

EINECSNumber : 204-146-8

RTECSNumber : AG6625000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only