missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl dichloroacetate, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 148625000

Catalog No. AC148625000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Methyl dichloroacetate
116-54-1
141°C to 145°C
Authentic
Glass bottle
1.4411 to 1.4431
MFCD00000843
02, 203
methyl dichloroacetate, dichloroacetic acid methyl ester, acetic acid, dichloro-, methyl ester, ccris 7740, acetic acid, 2,2-dichloro-, methyl ester, methyldichloroethanoate, acmc-1brpm, dichloroacetic acid methyl, methyl2,2-dichloroacetate, wln: gygvo1
HKMLRUAPIDAGIE-UHFFFAOYSA-N
methyl 2,2-dichloroacetate
8315
99%
99+%
1.3810g/mL
80°C
98.5% min. (GC)
C3H4Cl2O2
Cl2CHCO2CH3
500mL
1.381
Solubility in water: slightly soluble
COC(=O)C(Cl)Cl
142.97
142.97
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 204-146-8

RTECSNumber : AG6625000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only