missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl Ether, Gas, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC S2428

Catalog No. S2428


Explore available promotions

Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Methyl ether115-10-61000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Methyl ether
2mL amber prescored glass ampule
0.1,99.9
Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Methanol-P&T
LCGLNKUTAGEVQW-UHFFFAOYSA-N
methoxymethane
8254
115-10-6,67-56-1
1 mL
0.1,99.9
Certified Reference Material
C2H6O
MFCD00008494
COC
46.07
CHEBI:28887

Chemical Identifiers

115-10-6
46.07
LCGLNKUTAGEVQW-UHFFFAOYSA-N
CHEBI:28887
COC
C2H6O
MFCD00008494
8254
methoxymethane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Highly flammable liquid and vapor. May be fatal if swallowed and enters airways. Causes skin irritation. May cause drowsiness or dizziness. Suspected of damaging fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure.

DOTInformation : Nonhazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only