missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl Ethyl Ketone (ACS), Fisher Chemical™

Supplier:  Thermo Fisher Scientific Formerly Fairlawn M2094

 View more versions of this product

Catalog No. M209-4



Explore available promotions

Description

Description

Specifications

Specifications

Methyl Ethyl Ketone
78-93-3
0.806g/cm³
4 L
Glass Bottle
0.0025% max.
0.0005mEq/g max.
MFCD00011648
0.42 mPa/s at 15°C
2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
CCC(C)=O
72.11
CHEBI:28398
≥99%
-87°C
Colorless
80°C
Pass Test
Liquid
≥99%
C4H8O
105mbar at 20°C
0.1% max.
ZWEHNKRNPOVVGH-UHFFFAOYSA-N
butan-2-one
6569
72.11
Certified ACS Reagent

Chemical Identifiers

78-93-3
72.11
ZWEHNKRNPOVVGH-UHFFFAOYSA-N
6569
butan-2-one
C4H8O
MFCD00011648
2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
CHEBI:28398
CCC(C)=O
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Vapors may cause drowsiness and dizziness. May cause irritation of respiratory tract. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:3
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid

SDS Documents

Documents

Accessories
Promotions

Promotions