missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl hexanoate, 99%, Thermo Scientific Chemicals

$52.85 - $515.47

Chemical Identifiers

CAS 106-70-7
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00009510
InChI Key NUKZAGXMHTUAFE-UHFFFAOYSA-N
Synonym methyl caproate, hexanoic acid methyl ester, hexanoic acid, methyl ester, methyl hexoate, methyl capronate, methyl n-hexanoate, methyl hexylate, caproic acid methyl ester, methyl caproate natural, methyl n-hexoate
PubChem CID 7824
ChEBI CHEBI:77322
IUPAC Name methyl hexanoate
SMILES CCCCCC(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1075718
View Documents
Thermo Scientific Chemicals
A1075718
50 g
Each for $52.85
 
AAA107570B
View Documents
Thermo Scientific Chemicals
A107570B
1000 g
Each for $515.47
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

106-70-7
130.19
NUKZAGXMHTUAFE-UHFFFAOYSA-N
7824
methyl hexanoate
C7H14O2
MFCD00009510
methyl caproate, hexanoic acid methyl ester, hexanoic acid, methyl ester, methyl hexoate, methyl capronate, methyl n-hexanoate, methyl hexylate, caproic acid methyl ester, methyl caproate natural, methyl n-hexoate
CHEBI:77322
CCCCCC(=O)OC

Specifications

106-70-7
0.885
45°C (113°F)
1.405
50 g
1744683
NUKZAGXMHTUAFE-UHFFFAOYSA-N
methyl hexanoate
7824
130.19
Methyl hexanoate
-71°C
150°C to 151°C
C7H14O2
MFCD00009510
UN3272
methyl caproate, hexanoic acid methyl ester, hexanoic acid, methyl ester, methyl hexoate, methyl capronate, methyl n-hexanoate, methyl hexylate, caproic acid methyl ester, methyl caproate natural, methyl n-hexoate
CCCCCC(=O)OC
130.19
CHEBI:77322
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

EINECSNumber : 203-425-1

RTECSNumber : MO8401400

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only