missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl iso-Butyl Ketone (Certified ACS), Fisher Chemical™

Supplier:  Fisher Chemical M2131

 View more versions of this product

Catalog No. M213-1

Explore available promotions

Description

Description

Specifications

Specifications

Methyl iso-Butyl Ketone
108-10-1
117°C
Pass Test
Liquid
≥98.5 %
C6H12O
19.9mmHg at 25°C
4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
CC(C)CC(=O)C
100.161
CHEBI:82344
≥98.5%
-84°C
0.80g/cm³
1 L
Glass Bottle
0.005% max.
0.002mEq/g max.
MFCD00008938
0.1% max.
NTIZESTWPVYFNL-UHFFFAOYSA-N
4-methylpentan-2-one
7909
100.16
Certified ACS Reagent

Chemical Identifiers

108-10-1
100.161
NTIZESTWPVYFNL-UHFFFAOYSA-N
7909
4-methylpentan-2-one
C6H12O
MFCD00008938
4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
CHEBI:82344
CC(C)CC(=O)C
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Harmful by inhalation. Irritating to eyes, respiratory system and skin. May cause central nervous system effects. Repeated exposure may cause skin dryness or cracking. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. If peroxide formation is suspected, do not open or move container. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:2
Flammability:3
Instability:1

DOTInformation : DOT Class 3, : Flammable Liquid

SDS Documents

Documents

Promotions

Promotions