missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl isopropyl carbonate, 97%, Thermo Scientific Chemicals
$60.34 - $600.00
Chemical Identifiers
CAS | 51729-83-0 |
---|---|
Molecular Formula | C5H10O3 |
Molecular Weight (g/mol) | 118.13 |
MDL Number | MFCD01075689 |
InChI Key | RCIJMMSZBQEWKW-UHFFFAOYSA-N |
Synonym | methyl isopropyl carbonate, isopropyl methyl carbonate, acmc-20akf7, ch3 c o och ch3 2, carbonic acid methylisopropyl ester |
PubChem CID | 7021530 |
IUPAC Name | methyl propan-2-yl carbonate |
SMILES | COC(=O)OC(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1531403
|
Thermo Scientific Chemicals
L1531403 |
1 g |
Each for $60.34
|
|
|||||
AAL1531406
|
Thermo Scientific Chemicals
L1531406 |
5 g |
Each for $172.05
|
|
|||||
AAL1531414
|
Thermo Scientific Chemicals
L1531414 |
25 g |
Each for $600.00
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
51729-83-0 | |
118.13 | |
RCIJMMSZBQEWKW-UHFFFAOYSA-N | |
7021530 | |
COC(=O)OC(C)C |
C5H10O3 | |
MFCD01075689 | |
methyl isopropyl carbonate, isopropyl methyl carbonate, acmc-20akf7, ch3 c o och ch3 2, carbonic acid methylisopropyl ester | |
methyl propan-2-yl carbonate |
Specifications
51729-83-0 | |
116°C to 118°C | |
C5H10O3 | |
MFCD01075689 | |
UN3272 | |
methyl isopropyl carbonate, isopropyl methyl carbonate, acmc-20akf7, ch3 c o och ch3 2, carbonic acid methylisopropyl ester | |
COC(=O)OC(C)C | |
118.13 | |
118.13 | |
Methyl isopropyl carbonate |
0.975 | |
27°C (80°F) | |
1.383 | |
1 g | |
1744735 | |
RCIJMMSZBQEWKW-UHFFFAOYSA-N | |
methyl propan-2-yl carbonate | |
7021530 | |
97% |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only