missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl isovalerate, 98%, Thermo Scientific Chemicals

$78.48 - $211.84

Chemical Identifiers

CAS 556-24-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00042866
InChI Key OQAGVSWESNCJJT-UHFFFAOYSA-N
Synonym methyl isovalerate, methyl isopentanoate, methyl 3-methylbutyrate, isovaleric acid, methyl ester, butanoic acid, 3-methyl-, methyl ester, methyl isovalerianate, methylisovalerate, isovaleric acid methyl ester, methyl isovalerate natural, fema no. 2753
PubChem CID 11160
IUPAC Name methyl 3-methylbutanoate
SMILES CC(C)CC(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0624118
View Documents
Thermo Scientific Chemicals
L0624118
50 g
Each for $78.48
 
AAL0624130
View Documents
Thermo Scientific Chemicals
L0624130
250 g
Each for $211.84
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

556-24-1
116.16
OQAGVSWESNCJJT-UHFFFAOYSA-N
11160
CC(C)CC(=O)OC
C6H12O2
MFCD00042866
methyl isovalerate, methyl isopentanoate, methyl 3-methylbutyrate, isovaleric acid, methyl ester, butanoic acid, 3-methyl-, methyl ester, methyl isovalerianate, methylisovalerate, isovaleric acid methyl ester, methyl isovalerate natural, fema no. 2753
methyl 3-methylbutanoate

Specifications

556-24-1
115°C to 117°C
Glass bottle
1.3936
50 g
1699922
methyl isovalerate, methyl isopentanoate, methyl 3-methylbutyrate, isovaleric acid, methyl ester, butanoic acid, 3-methyl-, methyl ester, methyl isovalerianate, methylisovalerate, isovaleric acid methyl ester, methyl isovalerate natural, fema no. 2753
CC(C)CC(=O)OC
116.16
116.17
Methyl isovalerate
0.88
16°C (60°F)
C6H12O2
MFCD00042866
UN2400
0.88
OQAGVSWESNCJJT-UHFFFAOYSA-N
methyl 3-methylbutanoate
11160
98%
Safety and Handling

Safety and Handling

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H225

DOTInformation : Hazard Class: 3; Packaging Group: II

EINECSNumber : 209-117-3

RTECSNumber : NY1510000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only