missing translation for 'onlineSavingsMsg'
Learn More

Methyl L-(-)-Lactate 98.0+%, TCI America™

$29.83 - $223.68

Chemical Identifiers

CAS 27871-49-4
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
MDL Number MFCD00064265
InChI Key LPEKGGXMPWTOCB-VKHMYHEASA-N
Synonym methyl s---lactate, methyl l-lactate, s-methyl lactate, --methyl l-lactate, s-lactic acid methyl ester, methyl 2s-2-hydroxypropanoate, --methyl lactate, methyl s-2-hydroxypropionate, propanoic acid, 2-hydroxy-, methyl ester, 2s, --lactic acid methyl ester
PubChem CID 94386
ChEBI CHEBI:83222
IUPAC Name methyl (2S)-2-hydroxypropanoate
SMILES COC(=O)[C@H](C)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
L016325G
View Documents
TCI America
L016325G
25 g
Each for $29.83
Only null left
 
L0163100G
View Documents
TCI America
L0163100G
100 g
Each for $76.43
Only null left
 
L0163500G
View Documents
TCI America
L0163500G
500 g
Each for $223.68
Only null left
 
Description

Description

Specifications

Chemical Identifiers

27871-49-4
104.11
LPEKGGXMPWTOCB-VKHMYHEASA-N
94386
methyl (2S)-2-hydroxypropanoate
C4H8O3
MFCD00064265
methyl s---lactate, methyl l-lactate, s-methyl lactate, --methyl l-lactate, s-lactic acid methyl ester, methyl 2s-2-hydroxypropanoate, --methyl lactate, methyl s-2-hydroxypropionate, propanoic acid, 2-hydroxy-, methyl ester, 2s, --lactic acid methyl ester
CHEBI:83222
COC(=O)[C@H](C)O

Specifications

27871-49-4
140°C
MFCD00064265
3272
LPEKGGXMPWTOCB-VKHMYHEASA-N
methyl (2S)-2-hydroxypropanoate
94386
104.11
Liquid
Colorless
C4H8O3
25 g
methyl s---lactate, methyl l-lactate, s-methyl lactate, --methyl l-lactate, s-lactic acid methyl ester, methyl 2s-2-hydroxypropanoate, --methyl lactate, methyl s-2-hydroxypropionate, propanoic acid, 2-hydroxy-, methyl ester, 2s, --lactic acid methyl ester
COC(=O)[C@H](C)O
104.11
CHEBI:83222
≥98.0% (GC)
Methyl L-(-)-Lactate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1370

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'