missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl levulinate, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04339722

 View more versions of this product

Catalog No. AA4339722


Explore available promotions

Description

Description

Methyl levulinate is mainly used as raw material on perfume.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methyl levulinate
1.05 g/mL
72°C (161.6°F)
C6H10O3
100 g
It is soluble in ethanol,ether,acetone and benzene.
CC(=O)CCC(=O)OC
130.143
130.14
Liquid
624-45-3
191°C
Pleasant
MFCD00017499
methyl levulinate, methyllevulinate, levulinic acid methyl ester, pentanoic acid, 4-oxo-, methyl ester, methyl 4-oxovalerate, levulinic acid, methyl ester, 4-oxovaleric acid methyl ester, 4-oxo-pentanoic acid methyl ester, methyl levulate, me ester
UAGJVSRUFNSIHR-UHFFFAOYSA-N
methyl 4-oxopentanoate
69354
≥99%

Chemical Identifiers

624-45-3
130.143
UAGJVSRUFNSIHR-UHFFFAOYSA-N
69354
CC(=O)CCC(=O)OC
C6H10O3
MFCD00017499
methyl levulinate, methyllevulinate, levulinic acid methyl ester, pentanoic acid, 4-oxo-, methyl ester, methyl 4-oxovalerate, levulinic acid, methyl ester, 4-oxovaleric acid methyl ester, 4-oxo-pentanoic acid methyl ester, methyl levulate, me ester
methyl 4-oxopentanoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 210-846-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only