missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl Methacrylate, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor M127420LT

 View more versions of this product

Catalog No. 18611708


Explore available promotions

Description

Description

Specifications

Specifications

80-62-6
Steel Can
Approx. 25 ppm
Reagent
1
20 L
0.99

Chemical Identifiers

80-62-6
Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.