missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl methacrylate, 99%, stabilized, Thermo Scientific Chemicals

Methyl methacrylate, 99%, C5H8O2, CAS Number-80-62-6, 150-76-5, acryester m, diakon, methacrylic acid methyl ester, methyl 2-methyl-2-propenoate, methyl 2-methylpropenoate, methyl-methacrylat, methylmethacrylate, methyl methacrylate, methyl methylacrylate, pegalan, 1L

Supplier:  Thermo Scientific Chemicals 127140010

 View more versions of this product

Catalog No. AC127140010



Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methyl methacrylate
80-62-6,150-76-5
-48.0°C
0.9300g/mL
8°C
98.5% min. (GC)
C5H8O2
H2C=C(CH3)CO2CH3
1 L
0.93
methyl methacrylate, methylmethacrylate, methacrylic acid methyl ester, methyl methylacrylate, methyl 2-methylpropenoate, pegalan, methyl-methacrylat, diakon, acryester m, methyl 2-methyl-2-propenoate
VVQNEPGJFQJSBK-UHFFFAOYSA-N
methyl 2-methylprop-2-enoate
10 to 110ppm MEHQ
0.6 mPa.s (20°C)
100.12
Crystalline Powder or Crystals
Stabilized
0.005 % max. (as methacrylic acid)
White to Yellow
100.0°C
Authentic
Glass bottle
1.4130 to 1.4150
MFCD00008587
02, II, 398
15, 6013
Solubility in water: 15.9g/L (20°C). Other solubilities: soluble in most common organic solvents such as me,tetrahydrofuran,esters,aromatic and chlorinated,ethanol,ether,acetone
CC(=C)C(=O)OC
100.12
6658
CHEBI:34840
99%

Chemical Identifiers

80-62-6
100.12
VVQNEPGJFQJSBK-UHFFFAOYSA-N
6658
methyl 2-methylprop-2-enoate
C5H8O2
MFCD00008587
methyl methacrylate, methylmethacrylate, methacrylic acid methyl ester, methyl methylacrylate, methyl 2-methylpropenoate, pegalan, methyl-methacrylat, diakon, acryester m, methyl 2-methyl-2-propenoate
CHEBI:34840
CC(=C)C(=O)OC
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Highly flammable liquid and vapor.

GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 201-297-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only