Learn More
Methyl methacrylate, 99%, stabilized, Thermo Scientific Chemicals
Methyl methacrylate, 99%, C5H8O2, CAS Number-80-62-6, 150-76-5, acryester m, diakon, methacrylic acid methyl ester, methyl 2-methyl-2-propenoate, methyl 2-methylpropenoate, methyl-methacrylat, methylmethacrylate, methyl methacrylate, methyl methylacrylate, pegalan, 1L
Supplier: Thermo Scientific Chemicals 127140025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl methacrylate | |
80-62-6,150-76-5 | |
-48.0°C | |
0.9300g/mL | |
8°C | |
98.5% min. (GC) | |
C5H8O2 | |
H2C=C(CH3)CO2CH3 | |
2.5 L | |
0.93 | |
methyl methacrylate, methylmethacrylate, methacrylic acid methyl ester, methyl methylacrylate, methyl 2-methylpropenoate, pegalan, methyl-methacrylat, diakon, acryester m, methyl 2-methyl-2-propenoate | |
VVQNEPGJFQJSBK-UHFFFAOYSA-N | |
methyl 2-methylprop-2-enoate | |
10 to 110ppm MEHQ | |
0.6 mPa.s (20°C) | |
100.12 | |
Crystalline Powder or Crystals |
Stabilized | |
0.005 % max. (as methacrylic acid) | |
White to Yellow | |
100.0°C | |
Authentic | |
Glass bottle | |
1.4130 to 1.4150 | |
MFCD00008587 | |
02, II, 398 | |
15, 6013 | |
Solubility in water: 15.9g/L (20°C). Other solubilities: soluble in most common organic solvents such as me,tetrahydrofuran,esters,aromatic and chlorinated,ethanol,ether,acetone | |
CC(=C)C(=O)OC | |
100.12 | |
6658 | |
CHEBI:34840 | |
99% |
Chemical Identifiers
80-62-6 | |
100.12 | |
VVQNEPGJFQJSBK-UHFFFAOYSA-N | |
6658 | |
methyl 2-methylprop-2-enoate |
C5H8O2 | |
MFCD00008587 | |
methyl methacrylate, methylmethacrylate, methacrylic acid methyl ester, methyl methylacrylate, methyl 2-methylpropenoate, pegalan, methyl-methacrylat, diakon, acryester m, methyl 2-methyl-2-propenoate | |
CHEBI:34840 | |
CC(=C)C(=O)OC |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Highly flammable liquid and vapor.
GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 201-297-1
RUO – Research Use Only