missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl N,N-Dimethylanthranilate 97.0+%, TCI America™

$43.44 - $102.70

Chemical Identifiers

CAS 10072-05-6
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00025858
InChI Key ZCNSBHAIPOWHJE-UHFFFAOYSA-N
Synonym methyl 2-dimethylamino benzoate, methyl n,n-dimethylanthranilate, unii-77lqp0681t, n,n-dimethylanthranilic acid methyl ester, benzoic acid, 2-dimethylamino-, methyl ester, acmc-1bs8u, anthranilic acid, n,n-dimethyl-, methyl ester, methyl dimethyl anthranilate, methyln,n-dimethylanthranilate, methyl 2-dimethylamino-benzoate
PubChem CID 82336
IUPAC Name methyl 2-(dimethylamino)benzoate
SMILES CN(C)C1=CC=CC=C1C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D067510G
View Documents
TCI America
D067510G
10 g
Each for $43.44
 
D067525G
View Documents
TCI America
D067525G
25 g
Each for $102.70
 
Description

Description

Specifications

Chemical Identifiers

10072-05-6
179.219
ZCNSBHAIPOWHJE-UHFFFAOYSA-N
82336
CN(C)C1=CC=CC=C1C(=O)OC
C10H13NO2
MFCD00025858
methyl 2-dimethylamino benzoate, methyl n,n-dimethylanthranilate, unii-77lqp0681t, n,n-dimethylanthranilic acid methyl ester, benzoic acid, 2-dimethylamino-, methyl ester, acmc-1bs8u, anthranilic acid, n,n-dimethyl-, methyl ester, methyl dimethyl anthranilate, methyln,n-dimethylanthranilate, methyl 2-dimethylamino-benzoate
methyl 2-(dimethylamino)benzoate

Specifications

10072-05-6
131°C
MFCD00025858
methyl 2-dimethylamino benzoate, methyl n,n-dimethylanthranilate, unii-77lqp0681t, n,n-dimethylanthranilic acid methyl ester, benzoic acid, 2-dimethylamino-, methyl ester, acmc-1bs8u, anthranilic acid, n,n-dimethyl-, methyl ester, methyl dimethyl anthranilate, methyln,n-dimethylanthranilate, methyl 2-dimethylamino-benzoate
CN(C)C1=CC=CC=C1C(=O)OC
179.219
179.22
Liquid
Yellow
C10H13NO2
10 g
ZCNSBHAIPOWHJE-UHFFFAOYSA-N
methyl 2-(dimethylamino)benzoate
82336
≥97.0% (GC,T)
Methyl N,N-Dimethylanthranilate
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'