missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl nicotinoylacetate, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 425490025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Methyl nicotinoylacetate | |
54950-20-8 | |
94% min. (HPLC) | |
C9H9NO3 | |
2.5g | |
Solubility in water: neglegible | |
COC(=O)CC(=O)C1=CC=CN=C1 | |
179.18 | |
179.17 |
95% | |
Authentic | |
Glass bottle | |
MFCD00216524 | |
methyl nicotinoylacetate, methyl 3-oxo-3-pyridin-3-yl propanoate, 3-oxo-3-pyridin-3-ylpropionic acid methyl ester, 3-pyridinepropanoic acid, .beta.-oxo-, methyl ester, methyl 3-oxo-3-3-pyridyl propanoate, methyl 3-pyridinecarbonyl acetate, methyl 2-nicotinoyl acetate, 3-pyridinepropanoic acid, beta-oxo-, methyl ester, methyl-3-oxo-3 3-pyridinyl propanoate | |
JUQKVXRLRKKRPL-UHFFFAOYSA-N | |
methyl 3-oxo-3-pyridin-3-ylpropanoate | |
108645 | |
95% |
Chemical Identifiers
54950-20-8 | |
179.18 | |
JUQKVXRLRKKRPL-UHFFFAOYSA-N | |
108645 | |
COC(=O)CC(=O)C1=CC=CN=C1 |
C9H9NO3 | |
MFCD00216524 | |
methyl nicotinoylacetate, methyl 3-oxo-3-pyridin-3-yl propanoate, 3-oxo-3-pyridin-3-ylpropionic acid methyl ester, 3-pyridinepropanoic acid, .beta.-oxo-, methyl ester, methyl 3-oxo-3-3-pyridyl propanoate, methyl 3-pyridinecarbonyl acetate, methyl 2-nicotinoyl acetate, 3-pyridinepropanoic acid, beta-oxo-, methyl ester, methyl-3-oxo-3 3-pyridinyl propanoate | |
methyl 3-oxo-3-pyridin-3-ylpropanoate |
RUO â Research Use Only