missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl Nonafluorobutyl Ether 98.0+%, TCI America™
$65.17 - $485.86
Chemical Identifiers
CAS | 163702-07-6 |
---|---|
Molecular Formula | C5H3F9O |
Molecular Weight (g/mol) | 250.064 |
MDL Number | MFCD00674093 |
InChI Key | OKIYQFLILPKULA-UHFFFAOYSA-N |
Synonym | Methyl Perfluorobutyl Ether, Nonafluorobutyl Methyl Ether, Perfluorobutyl Methyl Ether |
PubChem CID | 164514 |
IUPAC Name | 1,1,1,2,2,3,3,4,4-nonafluoro-4-methoxybutane |
SMILES | COC(C(C(C(F)(F)F)(F)F)(F)F)(F)F |
Chemical Identifiers
163702-07-6 | |
250.064 | |
OKIYQFLILPKULA-UHFFFAOYSA-N | |
164514 | |
COC(C(C(C(F)(F)F)(F)F)(F)F)(F)F |
C5H3F9O | |
MFCD00674093 | |
Methyl Perfluorobutyl Ether, Nonafluorobutyl Methyl Ether, Perfluorobutyl Methyl Ether | |
1,1,1,2,2,3,3,4,4-nonafluoro-4-methoxybutane |
Specifications
163702-07-6 | |
C5H3F9O | |
25 g | |
OKIYQFLILPKULA-UHFFFAOYSA-N | |
1,1,1,2,2,3,3,4,4-nonafluoro-4-methoxybutane | |
164514 | |
≥98.0% (GC) | |
Methyl Nonafluorobutyl Ether |
Colorless | |
MFCD00674093 | |
Methyl Perfluorobutyl Ether, Nonafluorobutyl Methyl Ether, Perfluorobutyl Methyl Ether | |
COC(C(C(C(F)(F)F)(F)F)(F)F)(F)F | |
250.064 | |
250.06 | |
Liquid |
Safety and Handling
TSCA : Yes