Learn More
Methyl o-toluate, 99%, Thermo Scientific Chemicals
$59.81 - $793.60
Chemical Identifiers
CAS | 89-71-4 |
---|---|
Molecular Formula | C9H10O2 |
Molecular Weight (g/mol) | 150.177 |
MDL Number | MFCD00008428 |
InChI Key | WVWZECQNFWFVFW-UHFFFAOYSA-N |
Synonym | methyl o-toluate, methyl orthotoluate, o-toluic acid, methyl ester, methyl o-methylbenzoate, methyl toluate, benzoic acid, 2-methyl-, methyl ester, 2-methylbenzoic acid methyl ester, o-toluic acid methyl ester, methyl-o-methyl benzoate, 2-methyl methylbenzoate |
PubChem CID | 33094 |
IUPAC Name | methyl 2-methylbenzoate |
SMILES | CC1=CC=CC=C1C(=O)OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1569514
|
Thermo Scientific Chemicals
A1569514 |
25 g |
N/A
|
|
|||||
AAA1569522
|
Thermo Scientific Chemicals
A1569522 |
100 g |
N/A
|
|
|||||
AAA1569536
|
Thermo Scientific Chemicals
A1569536 |
500 g |
N/A
|
|
|||||
Description
It has been used in the preparation of 2-(azidomethyl)benzoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt has been used in the preparation of 2-(azidomethyl)benzoic acid.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away.
Chemical Identifiers
89-71-4 | |
150.177 | |
WVWZECQNFWFVFW-UHFFFAOYSA-N | |
33094 | |
CC1=CC=CC=C1C(=O)OC |
C9H10O2 | |
MFCD00008428 | |
methyl o-toluate, methyl orthotoluate, o-toluic acid, methyl ester, methyl o-methylbenzoate, methyl toluate, benzoic acid, 2-methyl-, methyl ester, 2-methylbenzoic acid methyl ester, o-toluic acid methyl ester, methyl-o-methyl benzoate, 2-methyl methylbenzoate | |
methyl 2-methylbenzoate |
Specifications
89-71-4 | |
207°C to 208°C | |
C9H10O2 | |
MFCD00008428 | |
2041846 | |
Not miscible or difficult to mix in water. | |
CC1=CC=CC=C1C(=O)OC | |
150.177 | |
150.18 | |
Methyl o-toluate |
1.072 | |
82°C (179°F) | |
1.519 | |
25 g | |
methyl o-toluate, methyl orthotoluate, o-toluic acid, methyl ester, methyl o-methylbenzoate, methyl toluate, benzoic acid, 2-methyl-, methyl ester, 2-methylbenzoic acid methyl ester, o-toluic acid methyl ester, methyl-o-methyl benzoate, 2-methyl methylbenzoate | |
WVWZECQNFWFVFW-UHFFFAOYSA-N | |
methyl 2-methylbenzoate | |
33094 | |
99% |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 201-932-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only