missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl octanoate, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1099118

 View more versions of this product

Catalog No. AAA1099118

Explore available promotions

Description

Description

Methyl octanoate-ethanol mixtures constitute the biodiesel-bioethanol surrogate fuel and kinetics of its oxidation has been studied experimentally in a jet-stirred reactor. Deoxygenation of methyl octanoate over alumina-supported Pt has been studied in both the vapor phase in a flow reactor and in the liquid phase in a semi-batch reactor. It is also used to make other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl octanoate-ethanol mixtures constitute the biodiesel-bioethanol surrogate fuel and kinetics of its oxidation has been studied experimentally in a jet-stirred reactor. Deoxygenation of methyl octanoate over alumina-supported Pt has been studied in both the vapor phase in a flow reactor and in the liquid phase in a semi-batch reactor. It is also used to make other chemicals.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

Methyl octanoate
-40°C
193°C to 194°C
C9H18O2
MFCD00009551
1752270
Insoluble in water.
CCCCCCCC(=O)OC
158.241
CHEBI:87432
99%
111-11-5
0.875
72°C (161°F)
1.4175
50 g
methyl caprylate, caprylic acid methyl ester, methyl n-octanoate, octanoic acid, methyl ester, uniphat a20, octanoic acid methyl ester, methyl octylate, methyl caprylate natural, unii-7mo740x6ql, fema no. 2728
JGHZJRVDZXSNKQ-UHFFFAOYSA-N
methyl octanoate
8091
158.24

Chemical Identifiers

111-11-5
158.241
JGHZJRVDZXSNKQ-UHFFFAOYSA-N
8091
methyl octanoate
C9H18O2
MFCD00009551
methyl caprylate, caprylic acid methyl ester, methyl n-octanoate, octanoic acid, methyl ester, uniphat a20, octanoic acid methyl ester, methyl octylate, methyl caprylate natural, unii-7mo740x6ql, fema no. 2728
CHEBI:87432
CCCCCCCC(=O)OC
Videos
Safety and Handling

Safety and Handling

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 203-835-0

RTECSNumber : RH0778000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only