missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl octanoate, 99%
CAS: 111-11-5 | C9H18O2 | 158.24 g/mol
$98.47 - $1072.81
Chemical Identifiers
| CAS | 111-11-5 |
|---|---|
| Molecular Formula | C9H18O2 |
| Molecular Weight (g/mol) | 158.24 |
| MDL Number | MFCD00009551 |
| InChI Key | JGHZJRVDZXSNKQ-UHFFFAOYSA-N |
| Synonym | methyl caprylate, caprylic acid methyl ester, methyl n-octanoate, octanoic acid, methyl ester, uniphat a20, octanoic acid methyl ester, methyl octylate, methyl caprylate natural, unii-7mo740x6ql, fema no. 2728 |
| PubChem CID | 8091 |
| ChEBI | CHEBI:87432 |
| IUPAC Name | methyl octanoate |
| SMILES | CCCCCCCC(=O)OC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC414620250
|
Thermo Scientific Chemicals
414620250 |
25 g | Glass bottle |
Each for $98.47
|
|
||||
|
AC414621000
|
Thermo Scientific Chemicals
414621000 |
100 g | Glass bottle |
Each for $258.29
|
|
||||
|
AC414625000
|
Thermo Scientific Chemicals
414625000 |
500 g | Glass bottle |
Each for $1,072.81
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 111-11-5 | |
| 158.24 | |
| JGHZJRVDZXSNKQ-UHFFFAOYSA-N | |
| 8091 | |
| methyl octanoate |
| C9H18O2 | |
| MFCD00009551 | |
| methyl caprylate, caprylic acid methyl ester, methyl n-octanoate, octanoic acid, methyl ester, uniphat a20, octanoic acid methyl ester, methyl octylate, methyl caprylate natural, unii-7mo740x6ql, fema no. 2728 | |
| CHEBI:87432 | |
| CCCCCCCC(=O)OC |
Specifications
| 111-11-5 | |
| Colorless | |
| 79.0°C (12.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4165 to 1.4185 | |
| MFCD00009551 | |
| 0.878 | |
| Solubility in water: insoluble | |
| CCCCCCCC(=O)OC | |
| 158.24 | |
| CHEBI:87432 | |
| 99% | |
| Methyl octanoate, 99% |
| 1mg KOH/g max. | |
| 0.8780g/mL | |
| 73°C | |
| 98.5% min. (GC) | |
| C9H18O2 | |
| CH3(CH2)6COOCH3 | |
| 25 g | |
| methyl caprylate, caprylic acid methyl ester, methyl n-octanoate, octanoic acid, methyl ester, uniphat a20, octanoic acid methyl ester, methyl octylate, methyl caprylate natural, unii-7mo740x6ql, fema no. 2728 | |
| JGHZJRVDZXSNKQ-UHFFFAOYSA-N | |
| methyl octanoate | |
| 8091 | |
| 158.24 | |
| Liquid |
Safety and Handling
EINECSNumber : 203-835-0
RTECSNumber : RH0778000
TSCA : TSCA
RUO – Research Use Only