missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl Orange Indicator Solution, Modified (Methyl Orange-Xylene Cyanol Mixed Indicator), Ricca Chemical
$154.78 - $297.13
Specifications
Chemical Name or Material | Methyl Orange Indicator Solution |
---|---|
Name Note | (Methyl Orange-Xylene Cyanol Mixed Indicator) |
CAS | 547-58-0,2650-17-1,7732-18-5 |
Packaging | Amber Glass Bottle |
Color | Green |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
5030-4
|
Ricca Chemical Company
50304 |
120 mL | Amber Glass Bottle |
Each for $154.78
|
|
||||
5030-16
|
Ricca Chemical Company
503016 |
500 mL | Amber Glass Bottle |
Each for $209.44
|
|
||||
5030-32
|
Ricca Chemical Company
503032 |
1 L | Amber Glass Bottle |
Each for $297.13
|
|
||||
Specifications
Methyl Orange Indicator Solution | |
547-58-0,2650-17-1,7732-18-5 | |
Green |
(Methyl Orange-Xylene Cyanol Mixed Indicator) | |
Amber Glass Bottle | |
Liquid |