missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl p-tolyl sulfide, 97%, Thermo Scientific Chemicals

$67.58 - $204.53

Chemical Identifiers

CAS 623-13-2
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.23
MDL Number MFCD00008560
InChI Key VHILIAIEEYLJNA-UHFFFAOYSA-N
Synonym methyl p-tolyl sulfide, 4-methylthioanisole, 4-methylthio toluene, p-methylthio toluene, benzene, 1-methyl-4-methylthio, p-tolyl methyl sulfide, p-methylphenyl methyl sulfide, methyl p-cresyl sulfide, p-cresyl methyl sulfide
PubChem CID 69334
IUPAC Name 1-methyl-4-methylsulfanylbenzene
SMILES CSC1=CC=C(C)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0293306
View Documents
Thermo Scientific Chemicals
L0293306
5 g
Each for $67.58
 
AAL0293314
View Documents
Thermo Scientific Chemicals
L0293314
25 g
Each for $204.53
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

623-13-2
138.23
VHILIAIEEYLJNA-UHFFFAOYSA-N
69334
CSC1=CC=C(C)C=C1
C8H10S
MFCD00008560
methyl p-tolyl sulfide, 4-methylthioanisole, 4-methylthio toluene, p-methylthio toluene, benzene, 1-methyl-4-methylthio, p-tolyl methyl sulfide, p-methylphenyl methyl sulfide, methyl p-cresyl sulfide, p-cresyl methyl sulfide
1-methyl-4-methylsulfanylbenzene

Specifications

623-13-2
81°C to 82°C (18 mmHg)
Unpleasant
1.573
5 g
1905733
VHILIAIEEYLJNA-UHFFFAOYSA-N
1-methyl-4-methylsulfanylbenzene
69334
97%
1.029
85°C (185°F)
C8H10S
MFCD00008560
UN3334
methyl p-tolyl sulfide, 4-methylthioanisole, 4-methylthio toluene, p-methylthio toluene, benzene, 1-methyl-4-methylthio, p-tolyl methyl sulfide, p-methylphenyl methyl sulfide, methyl p-cresyl sulfide, p-cresyl methyl sulfide
CSC1=CC=C(C)C=C1
138.23
138.23
Methyl p-tolyl sulfide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED LIQUID, N.O.S.

EINECSNumber : 210-773-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only