missing translation for 'onlineSavingsMsg'
Learn More

Methyl palmitate, 95%

CAS: 112-39-0 | C17H34O2 | 270.45 g/mol

$48.79 - $159.33

Chemical Identifiers

CAS 112-39-0
Molecular Formula C17H34O2
Molecular Weight (g/mol) 270.45
MDL Number MFCD00008994
InChI Key FLIACVVOZYBSBS-UHFFFAOYSA-N
Synonym methyl palmitate, palmitic acid methyl ester, hexadecanoic acid, methyl ester, palmitic acid, methyl ester, uniphat a60, metholene 2216, methyl n-hexadecanoate, hexadecanoic acid methyl ester, n-hexadecanoic acid methyl ester, unii-dpy8vcm98i
PubChem CID 8181
ChEBI CHEBI:69187
IUPAC Name methyl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC414681000
SDS View Documents
Thermo Scientific Chemicals
414681000
100 g Glass bottle
Each for $48.79
Only null left
 
AC414685000
SDS View Documents
Thermo Scientific Chemicals
414685000
500 g Glass bottle
Each for $159.33
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

112-39-0
270.45
FLIACVVOZYBSBS-UHFFFAOYSA-N
8181
methyl hexadecanoate
C17H34O2
MFCD00008994
methyl palmitate, palmitic acid methyl ester, hexadecanoic acid, methyl ester, palmitic acid, methyl ester, uniphat a60, metholene 2216, methyl n-hexadecanoate, hexadecanoic acid methyl ester, n-hexadecanoic acid methyl ester, unii-dpy8vcm98i
CHEBI:69187
CCCCCCCCCCCCCCCC(=O)OC

Specifications

112-39-0
Colorless
338.0°C
Authentic
Glass bottle
CH3(CH2)14CO2CH3
100 g
Solubility in water: insoluble. Other solubilities: solubel in alcohol,ether
CCCCCCCCCCCCCCCC(=O)OC
270.45
CHEBI:69187
95%
Methyl palmitate, 95%
28.0°C
0.85g/mL
>110°C
94% min. (GC)
C17H34O2
MFCD00008994
methyl palmitate, palmitic acid methyl ester, hexadecanoic acid, methyl ester, palmitic acid, methyl ester, uniphat a60, metholene 2216, methyl n-hexadecanoate, hexadecanoic acid methyl ester, n-hexadecanoic acid methyl ester, unii-dpy8vcm98i
FLIACVVOZYBSBS-UHFFFAOYSA-N
methyl hexadecanoate
8181
270.45
Liquid or Low Melting Solid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-966-3

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only