missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Phenoxyacetate 99.0+%, TCI America™
$223.68 - $2242.46
Chemical Identifiers
| CAS | 2065-23-8 |
|---|---|
| Molecular Formula | C9H10O3 |
| Molecular Weight (g/mol) | 166.176 |
| MDL Number | MFCD00010227 |
| InChI Key | BZCKRPHEZOHHBK-UHFFFAOYSA-N |
| Synonym | methyl phenoxyacetate, phenoxyacetic acid methyl ester, acetic acid, phenoxy-, methyl ester, methylphenoxyacetate, unii-jd9of3328o, phenoxy methyl acetate, methyl phenoxy acetate, phenoxyacetic acid methyl, acetic acid, 2-phenoxy-, methyl ester, pubchem12489 |
| PubChem CID | 16365 |
| IUPAC Name | methyl 2-phenoxyacetate |
| SMILES | COC(=O)COC1=CC=CC=C1 |
Chemical Identifiers
| 2065-23-8 | |
| 166.176 | |
| BZCKRPHEZOHHBK-UHFFFAOYSA-N | |
| 16365 | |
| COC(=O)COC1=CC=CC=C1 |
| C9H10O3 | |
| MFCD00010227 | |
| methyl phenoxyacetate, phenoxyacetic acid methyl ester, acetic acid, phenoxy-, methyl ester, methylphenoxyacetate, unii-jd9of3328o, phenoxy methyl acetate, methyl phenoxy acetate, phenoxyacetic acid methyl, acetic acid, 2-phenoxy-, methyl ester, pubchem12489 | |
| methyl 2-phenoxyacetate |
Specifications
| 2065-23-8 | |
| 245°C | |
| MFCD00010227 | |
| methyl phenoxyacetate, phenoxyacetic acid methyl ester, acetic acid, phenoxy-, methyl ester, methylphenoxyacetate, unii-jd9of3328o, phenoxy methyl acetate, methyl phenoxy acetate, phenoxyacetic acid methyl, acetic acid, 2-phenoxy-, methyl ester, pubchem12489 | |
| COC(=O)COC1=CC=CC=C1 | |
| 166.176 | |
| 166.18 | |
| Liquid |
| Yellow | |
| C9H10O3 | |
| 25 g | |
| BZCKRPHEZOHHBK-UHFFFAOYSA-N | |
| methyl 2-phenoxyacetate | |
| 16365 | |
| ≥99.0% (GC) | |
| Methyl Phenoxyacetate |
Safety and Handling
TSCA : Yes