missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl phenyl sulfoxide, 98+%, Thermo Scientific™

Chemical Identifiers

CAS 1193-82-4
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.21
MDL Number MFCD00002088
InChI Key JXTGICXCHWMCPM-UHFFFAOYSA-N
Synonym methyl phenyl sulfoxide, methylsulfinyl benzene, thioanisole s-oxide, phenyl methyl sulfoxide, sulfoxide, methyl phenyl, benzene, methylsulfinyl, methanesulfinylbenzene, methylphenylsulfoxide, methylsulphinyl benzene
PubChem CID 14516
IUPAC Name methylsulfinylbenzene
SMILES CS(=O)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC352120050
SDS View Documents
Thermo Scientific Chemicals
352120050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC352120250
SDS View Documents
Thermo Scientific Chemicals
352120250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1193-82-4
140.21
JXTGICXCHWMCPM-UHFFFAOYSA-N
14516
CS(=O)C1=CC=CC=C1
C7H8OS
MFCD00002088
methyl phenyl sulfoxide, methylsulfinyl benzene, thioanisole s-oxide, phenyl methyl sulfoxide, sulfoxide, methyl phenyl, benzene, methylsulfinyl, methanesulfinylbenzene, methylphenylsulfoxide, methylsulphinyl benzene
methylsulfinylbenzene

Specifications

1193-82-4
86°C
98% min. (GC)
C7H8OS
C6H5SOCH3
5g
Solubility in water: insoluble (20°C)
CS(=O)C1=CC=CC=C1
140.21
140.21
Methyl phenyl sulfoxide, 98%
92°C to 94°C (0.3mmHg)
Authentic
Glass bottle
1.5775
MFCD00002088
methyl phenyl sulfoxide, methylsulfinyl benzene, thioanisole s-oxide, phenyl methyl sulfoxide, sulfoxide, methyl phenyl, benzene, methylsulfinyl, methanesulfinylbenzene, methylphenylsulfoxide, methylsulphinyl benzene
JXTGICXCHWMCPM-UHFFFAOYSA-N
methylsulfinylbenzene
14516
98+%
Videos
Safety and Handling

Safety and Handling

MOISTURE SENSITIVE

HYGROSCOPIC

EINECSNumber : 214-781-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only