missing translation for 'onlineSavingsMsg'
Learn More

Methyl Red Indicator, 0.1% (w/v), Ricca Chemical

Supplier:  Ricca Chemical Company RI50604

 View more versions of this product

Catalog No. 5060-4


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Methyl Red Indicator
0.1% (w/v)
Indicator
Aqueous Solution
5
Liquid
845-10-3,7732-18-5
0.1% W/V
Natural Poly Bottle
Orange to Red
120 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions