Learn More
Methyl (S)-(+)-3-hydroxy-2-methylpropionate, 98%
CAS: 80657-57-4 | C5H10O3 | 118.13 g/mol
$234.78 - $1032.70
Chemical Identifiers
| CAS | 80657-57-4 |
|---|---|
| Molecular Formula | C5H10O3 |
| Molecular Weight (g/mol) | 118.13 |
| MDL Number | MFCD00064520 |
| InChI Key | ATCCIZURPPEVIZ-UHFFFAOYNA-N |
| Synonym | s-methyl 3-hydroxy-2-methylpropanoate, methyl s-+-3-hydroxy-2-methylpropionate, s-+-3-hydroxy-2-methylpropionic acid methyl ester, methyl 2s-3-hydroxy-2-methylpropanoate, 2-methyl-3-hydroxypropanoic acid, propanoic acid, 3-hydroxy-2-methyl-, methyl ester, 2s, s-+-methyl 3-hydroxy-2-methylpropionate, s-3-hydroxy-2-methylpropionic acid methyl ester, +-methyl l-beta-hydroxyisobutyrate, methyl 3-hydroxy-2-methylpropanoate # |
| PubChem CID | 10192563 |
| IUPAC Name | methyl (2S)-3-hydroxy-2-methylpropanoate |
| SMILES | COC(=O)C(C)CO |
Description
Methyl (S)-(+)-3-hydroxy-2-methylpropionate is used as a starting material for the synthesis of dictyostatin, discodermolide and spongidepsin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 80657-57-4 | |
| 118.13 | |
| ATCCIZURPPEVIZ-UHFFFAOYNA-N | |
| 10192563 | |
| COC(=O)C(C)CO |
| C5H10O3 | |
| MFCD00064520 | |
| s-methyl 3-hydroxy-2-methylpropanoate, methyl s-+-3-hydroxy-2-methylpropionate, s-+-3-hydroxy-2-methylpropionic acid methyl ester, methyl 2s-3-hydroxy-2-methylpropanoate, 2-methyl-3-hydroxypropanoic acid, propanoic acid, 3-hydroxy-2-methyl-, methyl ester, 2s, s-+-methyl 3-hydroxy-2-methylpropionate, s-3-hydroxy-2-methylpropionic acid methyl ester, +-methyl l-beta-hydroxyisobutyrate, methyl 3-hydroxy-2-methylpropanoate # | |
| methyl (2S)-3-hydroxy-2-methylpropanoate |
Specifications
| 80657-57-4 | |
| 1.07 g/mL | |
| 81°C (178°F) | |
| 1.425 | |
| 2 g | |
| s-methyl 3-hydroxy-2-methylpropanoate, methyl s-+-3-hydroxy-2-methylpropionate, s-+-3-hydroxy-2-methylpropionic acid methyl ester, methyl 2s-3-hydroxy-2-methylpropanoate, 2-methyl-3-hydroxypropanoic acid, propanoic acid, 3-hydroxy-2-methyl-, methyl ester, 2s, s-+-methyl 3-hydroxy-2-methylpropionate, s-3-hydroxy-2-methylpropionic acid methyl ester, +-methyl l-beta-hydroxyisobutyrate, methyl 3-hydroxy-2-methylpropanoate # | |
| ATCCIZURPPEVIZ-UHFFFAOYNA-N | |
| COC(=O)C(C)CO | |
| 118.13 | |
| 118.13 | |
| Liquid |
| Colorless | |
| 74°C (10 mmHg) | |
| C5H10O3 | |
| MFCD00064520 | |
| 1720830 | |
| Insoluble in water. Soluble in methanol. | |
| +27° (c=4 in methanol) | |
| methyl (2S)-3-hydroxy-2-methylpropanoate | |
| 10192563 | |
| 99% | |
| Methyl (S)- (+)-3-hydroxy-2-methylpropionate |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only