missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl stearate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1326514
Description
Methyl stearate is used as a nonionic surfactant, thereby enhancing the solubility of chemicals by dissociating aggregates and unfolding proteins. It is a fatty acid ester, which is used as an emulsifier and stabilizer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl stearate is used as a nonionic surfactant, thereby enhancing the solubility of chemicals by dissociating aggregates and unfolding proteins. It is a fatty acid ester, which is used as an emulsifier and stabilizer.
Solubility
Soluble in chloroform. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Methyl Stearate | |
38°C to 42°C | |
>110°C (230°F) | |
C19H38O2 | |
25 g | |
methyl stearate, octadecanoic acid, methyl ester, stearic acid methyl ester, metholene 2218, kemester 9718, methyl n-octadecanoate, stearic acid, methyl ester, kemester 9018, emery 2218, n-octadecanoic acid methyl ester | |
HPEUJPJOZXNMSJ-UHFFFAOYSA-N | |
methyl octadecanoate | |
8201 | |
298.51 |
112-61-8 | |
180°C to 182 (4 mm) | |
99% | |
MFCD00009005 | |
1786213 | |
Soluble in chloroform. Insoluble in water. | |
CCCCCCCCCCCCCCCCCC(=O)OC | |
298.511 | |
CHEBI:69188 |
Chemical Identifiers
112-61-8 | |
298.511 | |
HPEUJPJOZXNMSJ-UHFFFAOYSA-N | |
8201 | |
methyl octadecanoate |
C19H38O2 | |
MFCD00009005 | |
methyl stearate, octadecanoic acid, methyl ester, stearic acid methyl ester, metholene 2218, kemester 9718, methyl n-octadecanoate, stearic acid, methyl ester, kemester 9018, emery 2218, n-octadecanoic acid methyl ester | |
CHEBI:69188 | |
CCCCCCCCCCCCCCCCCC(=O)OC |
Safety and Handling
EINECSNumber : 203-990-4
RTECSNumber : WI4460000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only