missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Succinamate 98.0+%, TCI America™
$87.60 - $255.38
Chemical Identifiers
| CAS | 53171-39-4 |
|---|---|
| Molecular Formula | C5H9NO3 |
| Molecular Weight (g/mol) | 131.131 |
| InChI Key | HURZMSZDVGMYKJ-UHFFFAOYSA-N |
| Synonym | Succinamic Acid Methyl Ester |
| PubChem CID | 12550576 |
| IUPAC Name | methyl 4-amino-4-oxobutanoate |
| SMILES | COC(=O)CCC(=O)N |
Chemical Identifiers
| 53171-39-4 | |
| 131.131 | |
| Succinamic Acid Methyl Ester | |
| methyl 4-amino-4-oxobutanoate |
| C5H9NO3 | |
| HURZMSZDVGMYKJ-UHFFFAOYSA-N | |
| 12550576 | |
| COC(=O)CCC(=O)N |
Specifications
| 53171-39-4 | |
| White | |
| C5H9NO3 | |
| Succinamic Acid Methyl Ester | |
| COC(=O)CCC(=O)N | |
| 131.131 | |
| 131.13 | |
| Crystalline Powder |
| 89°C | |
| 118°C | |
| 5 g | |
| HURZMSZDVGMYKJ-UHFFFAOYSA-N | |
| methyl 4-amino-4-oxobutanoate | |
| 12550576 | |
| ≥98.0% (GC) | |
| Methyl Succinamate |
Safety and Handling
TSCA : No