missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl succinyl chloride, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 108400050

Catalog No. AC108400050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Methyl succinyl chloride
1.2320g/mL
73°C
Glass bottle
1.4402
MFCD00000750
02, II, 553
methyl 4-chloro-4-oxobutyrate, methyl 3-chloroformyl propionate, 3-carbomethoxypropionyl chloride, methyl 4-chloro-4-oxo-butanoate, 3-carbomethoxy propionyl chloride, butanoic acid, 4-chloro-4-oxo-, methyl ester, methoxycarbonylpropionyl chloride, 4-chloro-4-oxo-butanoicacimethylester, succinic acid monomethyl ester chloride, 4-chloro-4-oxobutyric acid methyl ester
COC(=O)CCC(=O)Cl
150.56
150.56
1490-25-1
58°C to 65°C (3.0mmHg)
96% min. (GC)
C5H7ClO3
CH3O2CCH2CH2COCl
5g
1.232
SRXOJMOGPYFZKC-UHFFFAOYSA-N
methyl 4-chloro-4-oxobutanoate
73888
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 216-077-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only