missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl tert-butylacetate, 98%, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals L0700509

Catalog No. AAL0700509


Explore available promotions

Description

Description

Methyl tert-butylacetate is an oxygenated solvent used alone or in blends for coatings, industrial cleaning and surface treatment. It is used as an intermediate to manufacture pharmaceuticals, synthetic flavorings and other organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl tert-butylacetate is an oxygenated solvent used alone or in blends for coatings, industrial cleaning and surface treatment. It is used as an intermediate to manufacture pharmaceuticals, synthetic flavorings and other organic
Specifications

Specifications

Methyl tert-butylacetate
126°C to 128°C
C7H14O2
MFCD00043646
UN3272
methyl tert-butylacetate, methyl 3,3-dimethylbutyrate, butanoic acid, 3,3-dimethyl-, methyl ester, butyric acid, 3,3-dimethyl-, methyl ester, methyl t-butylacetate, acmc-1c3ge, methyl-tert-butyl acetate, 3,3-dimethylbutyric acid methyl ester, 3,3-dimethyl-butyric acid methyl ester
DXBOTVWRXLQVMG-UHFFFAOYSA-N
methyl 3,3-dimethylbutanoate
66298
98%
10250-48-3
24°C (75°F)
1.3995
10 g
1747895
Not miscible in water.
CC(C)(C)CC(=O)OC
130.187
130.19

Chemical Identifiers

10250-48-3
130.187
DXBOTVWRXLQVMG-UHFFFAOYSA-N
66298
CC(C)(C)CC(=O)OC
C7H14O2
MFCD00043646
methyl tert-butylacetate, methyl 3,3-dimethylbutyrate, butanoic acid, 3,3-dimethyl-, methyl ester, butyric acid, 3,3-dimethyl-, methyl ester, methyl t-butylacetate, acmc-1c3ge, methyl-tert-butyl acetate, 3,3-dimethylbutyric acid methyl ester, 3,3-dimethyl-butyric acid methyl ester
methyl 3,3-dimethylbutanoate
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

EINECSNumber : 233-590-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only