missing translation for 'onlineSavingsMsg'
Learn More

Methyl Thieno[3,2-b]thiophene-2-carboxylate 98.0+%, TCI America™

$246.05 - $740.03

Chemical Identifiers

CAS 98800-10-3
Molecular Formula C8H6O2S2
Molecular Weight (g/mol) 198.254
MDL Number MFCD01934498
InChI Key VCLNJFSCOKHCEJ-UHFFFAOYSA-N
Synonym Thieno[3,2-b]thiophene-2-carboxylic Acid Methyl Ester
PubChem CID 2739788
IUPAC Name methyl thieno[3,2-b]thiophene-5-carboxylate
SMILES COC(=O)C1=CC2=C(S1)C=CS2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M30011G
View Documents
TCI America
M30011G
1 g
Each for $246.05
Only null left
 
M30015G
View Documents
TCI America
M30015G
5 g
Each for $740.03
Only null left
 
Description

Description

Specifications

Chemical Identifiers

98800-10-3
198.254
VCLNJFSCOKHCEJ-UHFFFAOYSA-N
2739788
COC(=O)C1=CC2=C(S1)C=CS2
C8H6O2S2
MFCD01934498
Thieno[3,2-b]thiophene-2-carboxylic Acid Methyl Ester
methyl thieno[3,2-b]thiophene-5-carboxylate

Specifications

98800-10-3
Yellow
MFCD01934498
Thieno[3,2-b]thiophene-2-carboxylic Acid Methyl Ester
COC(=O)C1=CC2=C(S1)C=CS2
198.254
198.25
Crystalline Powder
98°C
C8H6O2S2
1 g
VCLNJFSCOKHCEJ-UHFFFAOYSA-N
methyl thieno[3,2-b]thiophene-5-carboxylate
2739788
≥98.0% (GC)
Methyl Thieno[3,2-b]thiophene-2-carboxylate
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents