missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl thiosalicylate, 97%, Thermo Scientific Chemicals

$131.50 - $131.50

Chemical Identifiers

CAS 4892-02-8
Molecular Formula C8H8O2S
Molecular Weight (g/mol) 168.22
MDL Number MFCD00060678
InChI Key BAQGCWNPCFABAY-UHFFFAOYSA-N
Synonym methyl thiosalicylate, methyl 2-mercaptobenzoate, methyl o-mercaptobenzoate, usaf ma-15, benzoic acid, 2-mercapto-, methyl ester, methyl-o-mercaptobenzoate, methyl-2-mercaptobenzoate, benzoic acid, o-mercapto-, methyl ester, 2-methoxycarbonyl thiophenol, 2-mercaptobenzoic acid methyl ester
PubChem CID 21009
IUPAC Name methyl 2-sulfanylbenzoate
SMILES COC(=O)C1=CC=CC=C1S
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC369600050
SDS View Documents
Thermo Scientific Chemicals
369600050
5 g Glass bottle
Each for $131.50
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4892-02-8
168.22
BAQGCWNPCFABAY-UHFFFAOYSA-N
21009
COC(=O)C1=CC=CC=C1S
C8H8O2S
MFCD00060678
methyl thiosalicylate, methyl 2-mercaptobenzoate, methyl o-mercaptobenzoate, usaf ma-15, benzoic acid, 2-mercapto-, methyl ester, methyl-o-mercaptobenzoate, methyl-2-mercaptobenzoate, benzoic acid, o-mercapto-, methyl ester, 2-methoxycarbonyl thiophenol, 2-mercaptobenzoic acid methyl ester
methyl 2-sulfanylbenzoate

Specifications

4892-02-8
1.2230g/mL
>110°C
96% min. (GC)
C8H8O2S
HSC6H4CO2CH3
5 g
1.223
BAQGCWNPCFABAY-UHFFFAOYSA-N
methyl 2-sulfanylbenzoate
21009
97%
Methyl thiosalicylate, 97%
Colorless to Yellow or Green
98.0°C to 100.0°C (2.0 mmHg)
Authentic
Glass bottle
1.5910 to 1.5930
MFCD00060678
10, 130
methyl thiosalicylate, methyl 2-mercaptobenzoate, methyl o-mercaptobenzoate, usaf ma-15, benzoic acid, 2-mercapto-, methyl ester, methyl-o-mercaptobenzoate, methyl-2-mercaptobenzoate, benzoic acid, o-mercapto-, methyl ester, 2-methoxycarbonyl thiophenol, 2-mercaptobenzoic acid methyl ester
COC(=O)C1=CC=CC=C1S
168.22
168.22
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only