missing translation for 'onlineSavingsMsg'
Learn More

Methyl trans-2-Octenoate 95.0+%, TCI America™

$227.41 - $680.38

Chemical Identifiers

CAS 7367-81-9
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.225
MDL Number MFCD00036620
InChI Key CJBQSBZJDJHMLF-BQYQJAHWSA-N
Synonym methyl trans-2-octenoate, methyl oct-2-enoate, methyl 2-octenoate, 2-octenoic acid, methyl ester, methyl e-oct-2-enoate, unii-9en9v40j78, 2-octenoic acid, methyl ester, e, 2-octenoic acid, methyl ester, 2e, 2-octenoic acid methyl ester, fema no. 3712
PubChem CID 5364532
IUPAC Name methyl (E)-oct-2-enoate
SMILES CCCCCC=CC(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M20145ML
View Documents
TCI America
M20145ML
5 mL
Each for $227.41
Only null left
 
M201425ML
View Documents
TCI America
M201425ML
25 mL
Each for $680.38
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7367-81-9
156.225
CJBQSBZJDJHMLF-BQYQJAHWSA-N
5364532
CCCCCC=CC(=O)OC
C9H16O2
MFCD00036620
methyl trans-2-octenoate, methyl oct-2-enoate, methyl 2-octenoate, 2-octenoic acid, methyl ester, methyl e-oct-2-enoate, unii-9en9v40j78, 2-octenoic acid, methyl ester, e, 2-octenoic acid, methyl ester, 2e, 2-octenoic acid methyl ester, fema no. 3712
methyl (E)-oct-2-enoate

Specifications

7367-81-9
91°C
MFCD00036620
methyl trans-2-octenoate, methyl oct-2-enoate, methyl 2-octenoate, 2-octenoic acid, methyl ester, methyl e-oct-2-enoate, unii-9en9v40j78, 2-octenoic acid, methyl ester, e, 2-octenoic acid, methyl ester, 2e, 2-octenoic acid methyl ester, fema no. 3712
CCCCCC=CC(=O)OC
156.225
156.23
Liquid
Colorless
C9H16O2
5 mL
CJBQSBZJDJHMLF-BQYQJAHWSA-N
methyl (E)-oct-2-enoate
5364532
≥95.0% (GC)
Methyl trans-2-Octenoate
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'