Learn More
Thermo Scientific Chemicals Methyl Violet 2B, pure, indicator grade
$100.51 - $100.51
Chemical Identifiers
CAS | 8004-87-3 |
---|---|
Molecular Formula | C24H28ClN3 |
MDL Number | MFCD00012568 |
Synonym | Basic Violet 1, C.I. 42535 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC228371000
|
Thermo Scientific Chemicals
228371000 |
100 g | Glass bottle |
Each for $100.51
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
8004-87-3 | |
C24H28ClN3 | |
MFCD00012568 | |
Basic Violet 1, C.I. 42535 |
Specifications
137.0°C | |
1 to 1.25 | |
MFCD00012568 | |
Basic Violet 1, C.I. 42535 | |
Authentic | |
10% max. (105°C) | |
Pure | |
75% min. (on dry substance) | |
100 g | |
Methyl violet 2B |
8004-87-3 | |
C24H28ClN3 | |
13, 755 | |
Solubility in water: soluble. Other solubilities: soluble in conc. sulfuric acid and nitric acid, soluble in chloroform, partially soluble in alcohol and glycerol | |
580 to 586nm (in 50:50 IMS:water) | |
393.95 | |
Glass bottle | |
Green | |
Crystalline Powder With Metal Luster |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECSNumber : GC8425000
TSCA : TSCA
RUO – Research Use Only