missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals Methyl Yellow, pure, indicator grade
$126.31 - $1584.29
Chemical Identifiers
CAS | 60-11-7 |
---|---|
Molecular Formula | C14H15N3 |
Molecular Weight (g/mol) | 225.30 |
MDL Number | MFCD00008308 |
InChI Key | JCYPECIVGRXBMO-UHFFFAOYSA-N |
Synonym | 4-Dimethylaminoazobenzene, N, N-Dimethyl-4-phenylazoaniline, Solvent Yellow 2 |
PubChem CID | 6053 |
ChEBI | CHEBI:17903 |
IUPAC Name | N,N-dimethyl-4-phenyldiazenylaniline |
SMILES | CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC151400250
|
Thermo Scientific Chemicals
151400250 |
25 g | Glass bottle |
Each for $126.31
|
|
||||
AC151401000
|
Thermo Scientific Chemicals
151401000 |
100 g | Glass bottle |
Each for $362.33
|
|
||||
AC151405000
|
Thermo Scientific Chemicals
151405000 |
500 g | Glass bottle |
Each for $1,584.29
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
60-11-7 | |
225.30 | |
JCYPECIVGRXBMO-UHFFFAOYSA-N | |
6053 | |
N,N-dimethyl-4-phenyldiazenylaniline |
C14H15N3 | |
MFCD00008308 | |
4-Dimethylaminoazobenzene, N, N-Dimethyl-4-phenylazoaniline, Solvent Yellow 2 | |
CHEBI:17903 | |
CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1 |
Specifications
111.0°C to 117.0°C | |
C14H15N3 | |
MFCD00008308 | |
Solubility in water: 13.6 mg/l. Other solubilities: soluble ca. 140g/L in acetone, soluble in benzene, chloroform, diethyl ether,, ethanol, petroleum ether, vegetable oils, alcohol,, ether, strong mineral acids and soils | |
Authentic | |
N,N-dimethyl-4-phenyldiazenylaniline | |
6053 | |
3% max. | |
Pure | |
25 g | |
Methyl Yellow |
60-11-7 | |
(CH3)2NC6H4N=NC6H5 | |
4-Dimethylaminoazobenzene, N, N-Dimethyl-4-phenylazoaniline, Solvent Yellow 2 | |
JCYPECIVGRXBMO-UHFFFAOYSA-N | |
CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1 | |
225.30 | |
CHEBI:17903 | |
225.29 | |
Glass bottle | |
Powder |
RUO – Research Use Only