missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylcyclohexane, 99%, extra pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 126580010

 View more versions of this product

Catalog No. AC126580010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methylcyclohexane
108-87-2
0.7700g/mL
−3°C
98.5% min. (GC)
C7H14
C6H11CH3
1 L
0.77
cyclohexane, methyl, hexahydrotoluene, cyclohexylmethane, toluene hexahydride, hexahydroxytoluene, sextone b, toluene, hexahydro, metylocykloheksan, methyl cyclohexane, methylcyclohexan
UAEPNZWRGJTJPN-UHFFFAOYSA-N
methylcyclohexane
7962
99%
Liquid
99%
-126.3°C
101.0°C
Authentic
Glass Bottle
1.4210 to 1.4230
MFCD00001497
05, 29
15, 6118
Solubility in water: 0.1g/L (20°C). Other solubilities: soluble in acetone,benzene,carbon tetrachloride,,ether,ethanol
CC1CCCCC1
98.19
98.19
Extra Pure

Chemical Identifiers

108-87-2
98.19
UAEPNZWRGJTJPN-UHFFFAOYSA-N
7962
CC1CCCCC1
C7H14
MFCD00001497
cyclohexane, methyl, hexahydrotoluene, cyclohexylmethane, toluene hexahydride, hexahydroxytoluene, sextone b, toluene, hexahydro, metylocykloheksan, methyl cyclohexane, methylcyclohexan
methylcyclohexane
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-624-3

RTECSNumber : GV6125000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only